    Sort by:
  • Search Within Results
GSK180736A
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   GSK180736A is a G protein-coupled receptor kinase 2 (GRK2) inhibitor with an IC50 of 0.77 μM.