    Sort by:
  • Search Within Results
Apatinib
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   Apatinib is an orally bioavailable, small-molecule receptor tyrosine kinase inhibitor with potential antiangiogenic and antineoplastic activities