    Sort by:
  • Search Within Results
AS-2444697 hydrochloride
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   AS-2444697 is a potent and selective interleukin-1 receptor-associated kinase 4 (IRAK4) inhibitor (IC₅₀ = 21 nM). It exhibits 30-fold selectivity...