You Searched For: SC-26196
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: BioVision
Description: SC-26196 is a selective Δ6 desaturase inhibitor (IC₅₀ = 0.2 μM in vitro. It displays selectivity over Δ5 and Δ9 desatura...