    Sort by:
  • Search Within Results
A37
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   A37 is a aldehyde dehydrogenase 1A1 (ALDH1A1) inhibitor (IC₅₀ = 4.6 μM; Ki = 300 nM). Displays selectivity for ALDH1A1 over eight other...