    Sort by:
  • Search Within Results
A-674563
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   A-674563 is a potent, orally available Akt1 inhibitor with an IC50 value of 11nM. Also displays activity against PKA and CDK2 with IC50 values of 16nM...