    Sort by:
  • Search Within Results
Bendamustine hydrochloride
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   Bendamustine is a novel alkylating agent with a benzimidazole ring giving it purine analogue-like properties.  Like other alkylating agents, bend...