Human MIP-1β (CCL4) Standard TMB ELISA Development Kit, PeproTech
Supplier: PeproTech
900-TM36 900-T36
10777-926KT 168.51 USD
10777-926 10771-670
Human MIP-1β (CCL4) Standard TMB ELISA Development Kit, PeproTech
Assays ELISAs
PeproTech's Human MIP-1β (CCL4) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human MIP-1β (CCL4) in a sandwich ELISA format.
  • Easy-to-use
  • Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human MIP-1β (CCL4) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human MIP-1β (CCL4) in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human MIP-1β (CCL4) in approximately 1000 ELISA plate wells.