Citraconic anhydride ≥98%, ProteoSpec™

Danger

Synonyms: 3-Methylfuran-2,5-dione

RMB14105-25C2
RCRMB1410525C2EA 126.91 USD
RCRMB1410525C2
Citraconic anhydride ≥98%, ProteoSpec™
Citraconic anhydride
Formula: C₅H₄O₃
MW: 112.08 g/mol
Boiling Pt: 213…214 °C
Melting Pt: 6…10 °C
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00005522
CAS Number: 616-02-4
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear, colorless
Assay (by GC) min 98 %
Conforms to Reference To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR