    Sort by:
  • Search Within Results
Bis(sulfosuccinimidyl) suberate sodium salt
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Ricca Chemical
Description:   ProteoSpec* BS3 Crosslinker Bis-sulfosuccinimidyl suberate, Molecular Weight: 572.43, Formula: C16H18N2Na2O14S2, Appearance White solid, Absolute Quantitative NMR min 90 %, Storage: 2 deg C - 8 deg C, Size: 50 mg