Fiber-optic Macintosh Laryngoscope Blades, Sklar®
Supplier: Sklar
07-1274 07-1268 07-1273 07-1269 07-1272
76193-064EA 293.12 USD
76193-064 76193-082 76193-062 76193-080 76191-592
Fiber-optic Macintosh Laryngoscope Blades, Sklar®
Scalpels and Scalpel Assemblies Scalpel Blades
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Macintosh Laryngoscope Blades are used for direct, rigid laryngoscopy procedures to view vocal cords, remove foreign objects from the throat, biopsy, or tracheal intubation. These fiber-optic blades follow the Green System of fiber-optic compatibility, which is indicated by a green bar where the blade attaches to a fiber-optic handle.