Surgical Blades #21, Sklar®
06-3060
76191-690BX 97.54 USD
76191-690
Surgical Blades #21, Sklar®
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
The disposable, nonsterile, stainless steel Surgical Blade #21 is a larger version on the #10 Scalpel Blade. It has a curved cutting edge and a flat, unsharpened back edge. The #21 blade is compatible with the #4 series and larger handles. This blade is one of the most traditional, commonly used blades and often used for making small incisions in skin and muscle.
Packaging: This blade is available in a box of 100.