Traceable® Sentry™ Thermometer, Control Company
Supplier: Control Company
90012-13
490012-954EA 96.06 USD
490012-954
Traceable® Sentry™ Thermometer, Control Company
The Traceable® Sentry™ thermometer is designed for simultaneous displays of high/low and current temperature.
  • High-impact ABS plastic case
  • Simple one-key reset operation resets minimum/maximum memories
  • Can be placed underwater, in refrigerator, or freezer
  • Unplugging the cable activates a sensor inside the unit for monitoring ambient temperature
Instantly view if freezer, water bath, or incubator is within temperature spec. Monitor temperature of any experiment when you’re not there overnight, weekends, even during lunch. Range is –58 to +158 °F. Resolution is 1° and accuracy is ±1 °C. Small cable diameter (¹/₁₆") allows refrigerator doors to close on it.
Certifications: Multi-point calibration on an individually-numbered Traceable® Certificate which assures accuracy from our ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA.
Delivery information: Supplied with battery, probe, 10-foot cable, wall mount, magnet, stand, hook and loop tape.