You Searched For: N-Oleoyldopamine
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (89165-494)
Supplier: Enzo Life Sciences
Description: Endogenous TRPV1 agonist (Ki=36nM, EC50=36nM) with weak affinity for rat CB1 receptor (Ki=1.6µM). Potent inhibitor of 5-lipoxygenase (IC50=7.5nM) and ...