You Searched For: Ansamitocin P-3
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (89165-000)
Supplier: Enzo Life Sciences
Description: Ansamitocin P-3 is a maytansine analog which displays potent cytotoxicity against the human solid tumor cell lines A-549 and HT-29. It shares the same...