    Sort by:
  • Search Within Results
AACOCF3
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Enzo Life Sciences
Description:   AACOCF3 is a trifluoromethylketone derivative of arachidonic acid. It is a selective inhibitor of human cytosolic PLA2 and Ca2+-independent PLA2 (iPLA...