    Sort by:
  • Search Within Results
Thioetheramide-phosphorylcholine
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Enzo Life Sciences
Description:   A potent substrate analog inhibitor of snake venom PLA2 (IC50=2 µM). It inhibits human recombinant sPLA2 (IC50=0.9 µM) selectively over cPLA2.