    Sort by:
  • Search Within Results
Cilastatin Sodium
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (89161-698)
Supplier:  Enzo Life Sciences
Description:   Cilastatin is a brush border dipeptidase inhibitor. It inhibits the conversion of leukotriene D4 to leukotriene E4. Nephroprotective in cyclosporine A...