    Sort by:
  • Search Within Results
Gabexate mesylate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Enzo Life Sciences
Description:   A competitive, reversible inhibitor of serine proteases tryptase (Ki=3.4nM), thrombin (Ki=500nM), uPA (Ki=1.3µM), plasmin (Ki=1.5µM), trypsin (Ki=2µM)...