    Sort by:
  • Search Within Results
Zardaverine
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Enzo Life Sciences
Description:   Selective, dual inhibitor of PDE III and PDE IV (IC50s=2.5 and 1.1 µM respectively). Anti-allergic activity, combined inhibition of PDE III and IV hav...