    Sort by:
  • Search Within Results
TACE substrate (fluorogenic)
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (89160-976)
Supplier:  Enzo Life Sciences
Description:   Fluorogenic substrate for TACE (ADAM17) (between Ala-Val), as well as MMP-3, MMP-7, MMP-12, and MMP-17, but not for ADAM19, MMP-13, nor MMP-14. Simila...