    Sort by:
You Searched For:
Kavain (+/-)
1  results were found
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Enzo Life Sciences
Description:   Kavain is one constituent of kava-kava, an herbal drug of the South Pacific islands, which displays anti-convulsive, anxiolytic, analgesic and muscle ...