    Sort by:
  • Search Within Results
SEW-2871
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (89160-556)
Supplier:  Enzo Life Sciences
Description:   SEW-2871 is a novel, potent EDG receptor agonist structurally unrelated to sphingosine-1-phosphate and is highly selective for the human S1P1 receptor...