    Sort by:
  • Search Within Results
ICRF-193
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Enzo Life Sciences
Description:   A topoisomerase II inhibitor that does not cause DNA damage. Induces apoptosis in several cell lines including K562 and Molt-4 cells. Induces a G2 che...