    Sort by:
  • Search Within Results
Mevastatin sodium
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (89158-600)
Supplier:  Enzo Life Sciences
Description:   This is the active carboxylate form of mevastatin. It is active in whole cells as well as in cell free assays.