    Sort by:
  • Search Within Results
N-Arachidonyl maleimide
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Enzo Life Sciences
Description:   NAM is a highly potent and irreversible inhibitor of MAG lipase (IC50=155 nM) which covalently binds to a cysteine residue located near the fatty acid...