You Searched For: SU4312
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Enzo Life Sciences
Description: Potent and selective inhibitor of VEGFR-2 (Flk-1, KDR). It displays a 100-fold greater potency for the unactivated form of the kinase (Ki=0.04µM) comp...