    Sort by:
HNMPA-(AM)3
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Enzo Life Sciences
Description:   HNMPA-(AM)3 is a cell-permeable analog of HNMPA. It inhibits insulin receptor tyrosine kinase activity (IC50=100 µM) and insulin-stimulated glucose ox...