    Sort by:
  • Search Within Results
Oleyloxyethyl phosphorylcholine
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Enzo Life Sciences
Description:   A potent and specific substrate analog PLA2 inhibitor (IC50=6.2 µM-porcine pancreatic PLA2). It inhibits human recombinant sPLA2 (IC50=13.7 µM) select...