    Sort by:
  • Search Within Results
N(5)-(1-Iminoethyl)-L-ornithine HCl
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Enzo Life Sciences
Description:   L-NIO is approximately 5 times more potent as an inhibitor of endothelial cell NO synthase than other arginine analogs such as L-NAME and L-NMMA. It i...