    Sort by:
  • Search Within Results
Rp-cAMPS
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (89157-014)
Supplier:  Enzo Life Sciences
Description:   Rp-cAMPS is a potent and specific competitive inhibitor of the activation of cAMP-dependent protein kinases by cAMP (IC50=4.9 µM) . Its cell permeabil...