    Sort by:
  • Search Within Results
CGP-37157
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Enzo Life Sciences
Description:   Potent, selective inhibitor of mitochondrial Na+Ca2+ exchange. IC50=0.4µM. Blocks mitochondrial permeability transition.