    Sort by:
  • Search Within Results
L-Mimosine
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (89156-898)
Supplier:  Enzo Life Sciences
Description:   A rare plant amino acid that reversibly blocks the cell cycle at G1-phase. Inhibits the hydroxylation of deoxyhypusine and thus inhibits eukaryote ini...