Wigmore Saw, OR-Grade, Sklar
Supplier: Sklar
11-1530
76191-836EA 101.44 USD
76191-836
Wigmore Saw, OR-Grade, Sklar
Saws
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
Sklar's® Wigmore cast saw has a solid handle with a serrated stainless steel blade. It is used to cut through plaster casts. The serrations offer an advantage for materials such as plaster.