Masterflex® L/S® High-Performance Precision Tygon E-Lab (E-3603) Pump Tubing, Cole Parmer
Supplier: Cole Parmer
06509-15 06509-36 06509-35 06509-24
76266-388PK 283.88 USD
76266-388 76266-394 76266-392 76266-390
Masterflex® L/S® High-Performance Precision Tygon E-Lab (E-3603) Pump Tubing, Cole Parmer
Tubing Transfer Tubing
Ideal for general transfer applications.
  • Inexpensive tubing for general laboratory applications
  • Clear for easy flow monitoring
  • Handles virtually all inorganic chemicals
  • Non-aging, non-oxidizing
  • Low gas permeability
  • Good for viscous fluids
  • High dielectric constant
Compatible with Standard and High-Performance pump heads.
Application suitability:
Acids Good
Alkalies Good
Organic solvents Not recommended
Pressure Good
Vacuum Good
Viscous fluids Excellent
Sterile fluids Poor
Suction lift for tubing sizes 15, 24, and 35 is 29 ft. H₂O (8.8 m H₂O). Suction lift for tubing size 36 is 27 ft. H₂O (8.3 m H₂O).
Certifications: Resin Certification for tubing size 15: FDA; NonDEHP; NonPhthalate; EUFood102011; RoHS; USP. Resin Certification for tubing sizes 24, 35, and 36: 19352004; ADCF; EUFood102011; FDA; NonDEHP; NonPhthalate; NSF51; RoHS; USP.