Masterflex® L/S® Precision Tygon E-Lab (E-3603) Pump Tubing, Cole Parmer
Supplier: Cole Parmer
06509-13 06509-14 06509-16 06509-17 06509-18 06509-25
76266-366PK 106.54 USD
76266-366 76266-364 76266-386 76266-384 76266-382 76266-380
Masterflex® L/S® Precision Tygon E-Lab (E-3603) Pump Tubing, Cole Parmer
Tubing Transfer Tubing
Ideal for general transfer applications.
  • Inexpensive tubing for general laboratory applications
  • Clear for easy flow monitoring
  • Handles virtually all inorganic chemicals
  • Non-aging, non-oxidizing
  • Low gas permeability
  • Good for viscous fluids
  • High dielectric constant
Compatible with Standard and Easy-Load® pump heads.
Application suitability:
Acids Good
Alkalies Good
Organic solvents Not recommended
Pressure Good
Vacuum Good
Viscous fluids Excellent
Sterile fluids Poor
Suction lift for tubing sizes 13, 14, 16, and 25 is 29 ft. H₂O (8.8 m H₂O). Suction lift for tubing sizes 17 and 18 is 22' H₂O (6.7 m H₂O).
Certifications: Resin Certification for tubing sizes 13, 14, 16, and 17: FDA; NonDEHP; NonPhthalate; EUFood102011; RoHS; USP. Resin Certification for tubing sizes 18 and 25: 19352004; ADCF; EUFood102011; FDA; NonDEHP; NonPhthalate; NSF51; RoHS; USP.