3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS) 10% in deionised water detergent, Sterile-Filtered

Supplier: BioVision
Warning

Synonyms: CHAPS

AB291109-1000ML AB291109-500
76265-152EA 548.96 USD
76265-152 76265-154
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS) 10% in deionised water detergent, Sterile-Filtered
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)

A 10% sterile-filtered solution in water of the zwitterionic detergent CHAPS (Cat. No. 1545).

Formula: C₃₂H₅₈N₂O₇S
MW: 614.89 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00081083
CAS Number: 75621-03-3
https://us.vwr.com/stibo/web/std.lang.all/29/02/23892902.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR