Heat-Shrink Tubing, Multi Colored Assortment, McMaster-Carr Supply
Supplier: McMaster-Carr
6334K41
MSPP-6334K41EA 119.46 USD
MSPP-6334K41
Heat-Shrink Tubing, Multi Colored Assortment, McMaster-Carr Supply
Tubing Heat-Shrink Tubing
Heat-shrink tubings are tight-fitting protective insulation for electrical connections and to protect wire and cable from the environment.
  • Made of polyolefin plastic
  • Highly flexible
  • Slide on type of installation
  • Temperature range: −65 to 275 °F
When heat is applied, the tubing contracts to conform the surroundings. To fit inside the tubing, the wire or cable assembly must be smaller than the tubings Int.Ø before shrinking. To have a tight fit, the smallest portion of the assembly must be the same as or larger than the I.Ø after shrinking.
The shrinking temperature of this tubings are measured as 215 °F and shrink ratio lies at the value of 2:1. This multi colored assortment contains clear, blue, green, red, white, yellow and black tubings.
Certifications: Blue, red, green, white, yellow and black tubings are meet AMS/MIL-DTL-23053/5 Class 1 specifications and clear tubing meets AMS/MIL-DTL-23053/5 Class 2 specifications.
Delivery information: The assortment includes 35 of 0.09", 28 of 0.13", 21 of 0.19" and 0.25" and 14 of 0.38" and 0.5" tubings. The values are denoting, I.Ø of the tubings before shrinking. Each size comes in seven colors.
Packaging: Packed in a compartmented plastic Box.