Apomorphine hydrochloride hemihydrate

Danger

Synonyms: (R)-5,6,6a,7-Tetrahydro-6- methyl-4H-dibenzo[de,g]quinoline-10,11-diol hydrochlorideR-(-)-6aβ-aporphine-10,11-diol hydrochloride

B1541-250 B1541-50
76265-444EA 87.57 USD
76265-444 76265-446
Apomorphine hydrochloride hemihydrate
Apomorphine hydrochloride hemihydrate

Apomorphine is an archetypical dopamine pan-receptor agonist. It displays anti-Parkinson’s activity in vivo. Protects against MPTP-induced neurotoxicity in a mouse model. In clinical use for Parkinson’s disease.

Formula: C₁₇H₁₇NO₂·HCl·1/2H₂O
MW: 312.79 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00150904
CAS Number: 41372-20-7
UN: 2811
ADR: ,III
https://us.vwr.com/stibo/web/std.lang.all/29/75/23892975.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR