2',3',5'-Tri-O-acetyluridine

Synonyms: Uridine triacetateUridine 2',3',5'-triacetate(2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)tetrahydrofuran-3,4-diyl diacetate

B1516-1G B1516-5G B1516-25G
76264-780EA 64.99 USD
76264-780 76264-784 76264-782
2',3',5'-Tri-O-acetyluridine
2',3',5'-Tri-O-acetyluridine

Uridine triacetate serves as an antidote for toxicities caused by 5-FU/capecitabine. Uridine triacetate delivers high concentrations of uridine, which competes with toxic 5-FU metabolites.

Formula: C₁₅H₁₈N₂O₉
MW: 370.31 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00023795
CAS Number: 4105-38-8
https://us.vwr.com/stibo/web/std.lang.all/29/50/23892950.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR