GNE-3511

Synonyms: 2-[[6-(3,3-difluoropyrrolidin-1-yl)-4-[1-(oxetan-3-yl)piperidin-4-yl]pyridin-2-yl]amino]pyridine-4-carbonitrile2-((6-(3,3-Difluoropyrrolidin-1-yl)-4-(1-(oxetan-3-yl)piperidin-4-yl)pyridin-2-yl)amino)isonicotinonitrile

B1341-5 B1341-25
76265-182EA 1225.87 USD
76265-182 76265-180
GNE-3511
GNE-3511

GNE-3511 is a potent and selective zipper kinase (DLK, MAP3K12) inhibitors.

Formula: C₂₃H₂₆F₂N₆O
MW: 440.49 g/mol
Storage Temperature: Freezer
MDL Number: MFCD28502215
CAS Number: 1496581-76-0
https://us.vwr.com/stibo/web/std.lang.all/29/31/23892931.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR