GDC0879

Synonyms: 2-[4-[(1E)-1-hydroxyimino-2,3-dihydroinden-5-yl]-3-pyridin-4-ylpyrazol-1-yl]ethanol

B1340-25 B1340-5
76265-176EA 227.66 USD
76265-176 76265-178
GDC0879
GDC0879

A novel, potent, and selective inhibitor of B-Raf with an IC50 value of 0.13 nM

Formula: C₁₉H₁₈N₄O₂
MW: 334.37 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16875916
CAS Number: 905281-76-7
https://us.vwr.com/stibo/web/std.lang.all/29/30/23892930.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR