NVP-AEW541

Synonyms: 7-[3-(azetidin-1-ylmethyl)cyclobutyl]-5-(3-phenylmethoxyphenyl)pyr rolo[2,3-d]pyrimidin-4-amine

B1335-25 B1335-5
76265-158EA 729.47 USD
76265-158 76265-416
NVP-AEW541
NVP-AEW541

NVP-AEW541 is a novel, potent and selective inhibitor of IGF-IR kinase.

Formula: C₂₇H₂₉N₅O
MW: 439.55 g/mol
Storage Temperature: Freezer
CAS Number: 475489-16-8
https://us.vwr.com/stibo/web/std.lang.all/29/18/23892918.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR