Columbianadin

Synonyms: 2-[(8S)-2-oxo-8,9-dihydrofuro[2,3-h]chromen-8-yl]propan-2-yl (Z)-2-methylbut-2-enoate

B1334-25 B1334-5
76265-412EA 87.57 USD
76265-412 76265-414
Columbianadin
Columbianadin

Columbianadin (CBN) is a coumarin-type compound and one of the main bioactive constituents of the underground part of the herb, Angelica pubescens. Columbianadin has calcium-channel blocking function, can inhibit depolarization induced Ca2+ uptake in rat pituitary GH3 cells.

Formula: C₁₉H₂₀O₅
MW: 328.36 g/mol
Storage Temperature: Freezer
MDL Number: MFCD07781426
CAS Number: 5058-13-9
https://us.vwr.com/stibo/web/std.lang.all/29/17/23892917.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR