BV6

Synonyms: (S,S,2S,2'S)-N,N'-((2S,2'S)-(hexane-1,6-diylbis(azanediyl))bis(3-oxo-1,1-diphenylpropane-3,2-diyl))bis(1-((S)-2-cyclohexyl-2-((S)-2-(methylamino)(2S,2'S)-N,N'-((2S,2'S)-(Hexane-1,6-diylbis(azanediyl))bis(1-oxo-3,3-diphenylpropane-1,2-diyl))bis(1-((S)-2-cyclohexyl-2-((S)-2-(methylamino)propanamido)acetyl)pyrrolidine-2-carboxamide)

B1332-5 B1332-25
76265-404EA 204.31 USD
76265-404 76265-406
BV6
BV6

BV6 is a selective inhibitor of IAP proteins

Formula: C₇₀H₉₆N₁₀O₈
MW: 1205.57 g/mol
Storage Temperature: Freezer
MDL Number: MFCD28168021
CAS Number: 1001600-56-1
https://us.vwr.com/stibo/web/std.lang.all/29/15/23892915.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR