BMX-IN-1

Synonyms: N-[5-[9-[4-(methanesulfonamido)phenyl]-2-oxobenzo[h][1,6]naphthyridin-1-yl]-2-methylphenyl]prop-2-enamideN-[5-[9-[4-(methanesulphonamido)phenyl]-2-oxobenzo[h][1,6]naphthyridin-1-yl]-2-methylphenyl]prop-2-enamide

B1330-5 B1330-25
76265-396EA 321.07 USD
76265-396 76265-398
BMX-IN-1
BMX-IN-1

BMX-IN-1 is a highly selective and potent irreversible inhibitor of BMX kinase.

Formula: C₂₉H₂₄N₄O₄S
MW: 524.59 g/mol
Storage Temperature: Freezer
CAS Number: 1431525-23-3
https://us.vwr.com/stibo/web/std.lang.all/29/12/23892912.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR