GNF-5

Synonyms: N-(2-hydroxyethyl)-3-[6-[4-(trifluoromethoxy)anilino]pyrimidin-4-yl]benzamideN-(2-Hydroxyethyl)-3-(6-((4-(trifluoromethoxy)phenyl)amino)pyrimidin-4-yl)benzamide

B1329-5 B1329-25
76265-392EA 99.24 USD
76265-392 76265-394
GNF-5
GNF-5

GNF-5 is a selective non-ATP competitive inhibitor of Bcr-Abl

Formula: C₂₀H₁₇F₃N₄O₃
MW: 418.37 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16877246
CAS Number: 778277-15-9
https://us.vwr.com/stibo/web/std.lang.all/29/11/23892911.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR