Entrectinib

Synonyms: N-(5-(3,5-Difluorobenzyl)-1H-indazol-3-yl)-4-(4-methylpiperazin-1-yl)-2-((tetrahydro-2H-pyran-4-yl)amino)benzamide(Z)-N-(5-(3,5-difluorobenzyl)-1H-indazol-3(2H)-ylidene)-4-(4-methylpiperazin-1-yl)-2-((tetrahydro-2H-pyran-4-yl)amino)benzamide

B1324-5 B1324-25
76265-372EA 145.89 USD
76265-372 76265-374
Entrectinib
Entrectinib

Entrectinib is an inhibitor of TrkA (IC₅₀ = 1.7 nM), TrkB (IC₅₀ = 0.1 nM), and TrkC (IC₅₀ = 0.1 nM), as well as C-ros oncogene 1 (ROS1; IC₅₀ = 0.2 nM) and anaplastic lymphoma kinase (ALK; IC₅₀ = 1.6 nM). Entrectinib blocks proliferation of ALK-dependent cell lines, including those with L1196M or C1156Y resistance mutations, and inhibits ALK‐dependent signaling

Formula: C₃₁H₃₄F₂N₆O₂
MW: 560.64 g/mol
Storage Temperature: Freezer
MDL Number: MFCD28129099
CAS Number: 1108743-60-7
https://us.vwr.com/stibo/web/std.lang.all/29/05/23892905.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR