Ear Tag Applicator, Kent Scientific
INS1005-1S
MSPP-INS10051SEA 70.77 USD
MSPP-INS10051S
Ear Tag Applicator, Kent Scientific
This hand-held ear tag applicator is made of stainless steel, is designed to seal the ear tags with minimal pressure, while providing a good, secure seal. The spring applies slight closing pressure to help hold the tag in the applicator prior to sealing.
  • Ear tag applicator can be used with or without the spring
Place the tag in the applicator keeping the hole side of the tag flat against the jaw of the applicator with the indentation. Locate the tag within the ear, applying beyond a ring of cartilage. After sealing, make sure that the tag point has come through the hole and bent around, sealing properly.