RNAse H
Supplier: BioVision
M1224-100
76264-950EA 286.16 USD
76264-950
RNAse H
Thermostable RNAse H (Ribonuclease H) is an endoribonuclease that specifically hydrolyzes the phosphodiester bonds of RNA strands in RNA-DNA hybrids.
  • High-stringency hybrid selection and mapping of mRNA structure
  • Removal of mRNA prior to synthesis of second strand cDNA
  • Removal of the poly(A) sequences from mRNA in the presence of oligo (dT)
  • Directed cleavage of RNA